Glutaraldehyde 25% aqueous solution
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Glutaral, Glutardialdehyde, 1,5-Pentanedial
A17876-AE
A17876-0F
A17876-AP
CAAAAA17876-AEEA
51.67
CAD
CAAAAA17876-AE
CAAAA17876-0F
CAAAA17876-AP
Glutaraldehyde 25% aqueous solution
Glutaraldehyde
CAS No.: 111-30-8
Molecular Formula: C5H8O2
Formula Weight: 100.12
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Density: 1.062
Refractive Index: 1.3730
Appearance: Colorless
Beilstein Reference: 605390
Merck Reference: 14,4472
UN No.: 2922
MDL No.: MFCD00007025
Formula:
C₅H₈O₂ MW: 100.12 Boiling Pt: 188 Melting Pt: -10 Density: 1.062 Flash Pt: >71 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007025 CAS Number: 111-30-8 EINECS: 203-856-5 UN: 3265 ADR: 8 III Merck Index: 14,4472 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...