DL-Malic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A17874-0E A17874-36
CAAAA17874-0EEA 155.63 CAD
CAAAA17874-0E CAAAA17874-36
DL-Malic acid 98%
DL-Malic acid

MDL: MFCD00064212 Beilstein: 1723539 Soluble in acetone, dioxane, water, methanol and ethanol. Insoluble in benzene.


Purity: 98% CAS No.: 6915-15-7 Molecular Formula: C4H6O5 Formula Weight: 134.09 Flash Point: 203°(397°F) Physical Form: Powder Density: 1.609 Appearance: White to pale cream Melting Point: 130-133° Solubility: Soluble in acetone, dioxane, water, methanol and ethanol. Insoluble in benzene. Beilstein Reference: 1723539 Merck Reference: 14,5707 MDL No.: MFCD00064212

Formula: HOOCCH(OH)CH₂COOH
Boiling Pt: 306 °C (1013 hPa)
Melting Pt: 130…133 °C
Density: 1.609
Flash Pt: 203 °C(397 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064212
CAS Number: 617-48-1
EINECS: 230-022-8
Merck Index: 14,5707
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=920b54632a869bc5df22624fb9fecd4d11cae65e&vid=278193678126274041defdc0bd71c7c40efb3085&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR