2-Chloro-m-xylene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Chloro-1,3-dimethylbenzene, 1-Chloro-2,6-dimethylbenzene
A17867-36
A17867-14
A17867-22
CAAAAA17867-36EA
1434.64
CAD
CAAAAA17867-36
CAAAA17867-14
CAAAA17867-22
2-Chloro-m-xylene 98%
2-Chloro-m-xylene
Purity: 98%
CAS No.: 6781-98-2
Molecular Formula: C8H9Cl
Formula Weight: 140.61
Flash Point: 63°(145°F)
Refractive Index: 1.5270
Melting Point: -36°
Boiling Point: 182-183°
Beilstein Reference: 2040954
MDL No.: MFCD00000565
Formula:
C₈H₉Cl MW: 140.61 g/mol Boiling Pt: 182…183 °C Melting Pt: –36 °C Density: 1.061 Flash Pt: 63 °C(145 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000565 CAS Number: 6781-98-2 EINECS: 229-843-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...