Tetramethylammonium fluoride tetrahydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

A17864-06 A17864-14 A17864-22
CAAAA17864-06EA 65.25 CAD
CAAAA17864-06 CAAAA17864-14 CAAAA17864-22
Tetramethylammonium fluoride tetrahydrate 98%
Tetramethylammonium fluoride tetrahydrate

Purity: 98% CAS No.: 17787-40-5 Molecular Formula: (CH3)4NF.4H2O Formula Weight: 165.21 (93.14anhy) Flash Point: 110°(230°F) Odor: Odorless Beilstein Reference: 3916184 UN No.: 2811 MDL No.: MFCD00149968

Formula: C₄H₁₂FN·4H₂O
MW: 165.21 g/mol
Melting Pt: 42…46 °C
Density: 1.115 g/cm³ (20 °C)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00149968
CAS Number: 17787-40-5
EINECS: 206-769-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR