Hexafluoroglutaric acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17848-06
A17848-14
CAAAA17848-06EA
96.06
CAD
CAAAA17848-06
CAAAA17848-14
Hexafluoroglutaric acid 98%
Hexafluoroglutaric acid
Purity: 98%
CAS No.: 376-73-8
Molecular Formula: C5H2F6O4
Formula Weight: 240.06
Melting Point: 92-97°
Boiling Point: 134-138°/3mm
UN No.: 3261
MDL No.: MFCD00004172
Formula:
C₅H₂F₆O₄ Boiling Pt: 134…138 °C (3 mmHg) Melting Pt: 92…97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004172 CAS Number: 376-73-8 EINECS: 206-813-9 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...