3-Phenoxypropionic acid 98+%

Supplier: Thermo Scientific Chemicals

A17842-03 A17842-06 A17842-14
CAAAA17842-03EA 34.6 CAD
CAAAA17842-03 CAAAA17842-06 CAAAA17842-14
3-Phenoxypropionic acid 98+%
3-Phenoxypropionic acid

Purity: 98+% CAS No.: 7170-38-9 Molecular Formula: C6H5O(CH2)2CO2H Formula Weight: 166.17 Melting Point: 96-99° Boiling Point: 243-245° MDL No.: MFCD00002769

Formula: C₉H₁₀O₃
Boiling Pt: 243…245 °C
Melting Pt: 96…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00002769
CAS Number: 7170-38-9
EINECS: 230-518-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR