N,N-Diethyl-p-phenylenediammonium sulfate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Amino-N,N-diethylaniline sulfate, N,N-Diethyl-p-phenylenediamine sulphate, N,N-Diethyl-1,4-phenylenediammonium sulphate, N1,N1-Diethylbenzene-1,4-diamine sulfate, 4-Amino-N,N-diethylaniline sulphate, N,N-Diethyl-1,4-phenylenediammonium sulfate, N,N-Diethyl-p-phenylenediamine sulfate, N,N-Diethylbenzene-1,4-diammonium sulfate, N1,N1-Diethylbenzene-1,4-diamine sulphate, N,N-Diethylbenzene-1,4-diammonium sulphate

A17832-0B A17832-18 A17832-30
CAAAA17832-0BEA 474 CAD
CAAAA17832-0B CAAAA17832-18 CAAAA17832-30
N,N-Diethyl-p-phenylenediammonium sulfate 97%
N,N-Diethyl-p-phenylenediammonium sulphate

Purity: 97% CAS No.: 6283-63-2 Molecular Formula: C10H16N2.H2SO4 Formula Weight: 262.33 Melting Point: 183-188° Beilstein Reference: 4219768

Formula: C₁₀H₁₈N₂O₄S
Boiling Pt: 274 - 275 °C (1013 hPa)
Melting Pt: 183…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00012993
CAS Number: 6283-63-2
EINECS: 228-500-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR