Dehydroacetic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17827-0B
A17827-18
A17827-30
CAAAA17827-0BEA
328.64
CAD
CAAAA17827-0B
CAAAA17827-18
CAAAA17827-30
Dehydroacetic acid 98%
Dehydracetic acid
Purity: 98%
CAS No.: 520-45-6
Molecular Formula: C8H8O4
Formula Weight: 168.15
Flash Point: 157°(314°F)
Melting Point: 109-113°
Boiling Point: 269-270°
Beilstein Reference: 6129
Merck Reference: 14,2865
MDL No.: MFCD00066709
Formula:
C₈H₈O₄ Boiling Pt: 269…270 °C Melting Pt: 109…113 °C Flash Pt: 157 °C(314 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00066709 CAS Number: 520-45-6 EINECS: 208-293-9 Merck Index: 14,2865 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...