(1S)-(-)-β-Pinene 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A17818-AE
A17818-0F
A17818-AP
CAAAAA17818-AEEA
49.45
CAD
CAAAAA17818-AE
CAAAA17818-0F
CAAAA17818-AP
(1S)-(-)-β-Pinene 99%
(1S)-(-)-β-Pinene
Purity: 99%
CAS No.: 18172-67-3
Molecular Formula: C10H16
Formula Weight: 136.24
Flash Point: 36°(96°F)
Refractive Index: 1.4780
Odor: Turpentine-like
Melting Point: -61°
Boiling Point: 164-165°
Beilstein Reference: 2038282
Merck Reference: 14,7446
Optical Rotation: -21° (neat)
UN No.: 2319
MDL No.: MFCD00001345
Formula:
C₁₀H₁₆ Boiling Pt: 164…165 °C Melting Pt: –61 °C Density: 0.860 Flash Pt: 36 °C(96 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001345 CAS Number: 18172-67-3 EINECS: 242-060-2 UN: 2319 ADR: "3",III Merck Index: 14,7446 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...