1,2,4,5-Tetrafluorobenzene 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A17815-06
A17815-14
CAAAA17815-06EA
59.41
CAD
CAAAA17815-06
CAAAA17815-14
1,2,4,5-Tetrafluorobenzene 99%
1,2,4,5-Tetrafluorobenzene
Purity: 99%
CAS No.: 327-54-8
Molecular Formula: C6H2F4
Formula Weight: 150.07
Flash Point: 16°(60°F)
Refractive Index: 1.4070
Melting Point: 3-5°
Boiling Point: 90°
Beilstein Reference: 1909052
UN No.: 2387
MDL No.: MFCD00000307
Formula:
C₆H₂F₄ MW: 150.08 g/mol Boiling Pt: 90 °C Melting Pt: 3…5 °C Density: 1.430 Flash Pt: 16 °C (60 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000307 CAS Number: 327-54-8 EINECS: 206-319-3 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...