AMP (2-Amino-2-methylpropanol), (max. 5% H₂O) 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17814-AE
A17814-0F
A17814-AP
CAAAAA17814-AEEA
43.45
CAD
CAAAAA17814-AE
CAAAA17814-0F
CAAAA17814-AP
AMP (2-Amino-2-methylpropanol), (max. 5% H₂O) 95%
AMP (2-Amino-2-methylpropanol)
Purity: 95%
CAS No.: 124-68-5
Molecular Formula: C4H11NO
Formula Weight: 89.14
Flash Point: 67°(152°F)
Refractive Index: 1.4455
Odor: Amine-like
pH: 11.3
Melting Point: 24-28°
Boiling Point: 164-166°
Beilstein Reference: 505979
Merck Reference: 14,449
MDL No.: MFCD00008051
Formula:
(CH₃)₂C(NH₂)CH₂OH Boiling Pt: 164…166 °C Melting Pt: 24…28 °C Density: 0.934 Flash Pt: 67 °C(152 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008051 CAS Number: 124-68-5 EINECS: 204-709-8 Merck Index: 14,449 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...