Methanesulfonic acid ≥70% in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

A17803-0F A17803-AE A17803-AP
CAAAA17803-0FEA 276.5 CAD
CAAAA17803-0F CAAAA17803-AE CAAAA17803-AP
Methanesulfonic acid ≥70% in aqueous solution
Methanesulphonic acid

CAS No.: 75-75-2 Molecular Formula: CH3SO3H Formula Weight: 96.10 Flash Point: 110°(230°F) Refractive Index: 1.4150 Merck Reference: 14,5954 MDL No.: MFCD00007518

Formula: CH₄O₃S
Density: 1.320
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007518
CAS Number: 75-75-2
EINECS: 200-898-6
UN: 3265
ADR: 8,III
Merck Index: 14,5954
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bebd3f86cbdbb894828247119a403c686b0ecddf&vid=01bb7db9690bd0b27a8ab4f277f8a301da72087f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR