2-Nitrochalcone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17763-06
A17763-14
CAAAA17763-06EA
95.43
CAD
CAAAA17763-06
CAAAA17763-14
2-Nitrochalcone 97%
2-Nitrochalcone
Purity: 97%
CAS No.: 7473-93-0
Molecular Formula: C15H11NO3
Formula Weight: 253.26
Melting Point: 124-128°
MDL No.: MFCD00031069
Formula:
C₁₅H₁₁NO₃ Melting Pt: 124…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00031069 CAS Number: 7473-93-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...