2-Chlorobenzothiazole 98%

Supplier: Thermo Scientific Chemicals
Warning

A17762-22 A17762-06 A17762-14
CAAAAA17762-22EA 497.7 CAD
CAAAAA17762-22 CAAAA17762-06 CAAAA17762-14
2-Chlorobenzothiazole 98%
2-Chlorobenzothiazole

Purity: 98+% CAS No.: 615-20-3 Molecular Formula: C7H4ClNS Formula Weight: 169.63 Flash Point: 110°(230°F) Refractive Index: 1.6370 Melting Point: 30-33° Boiling Point: 141°/30mm Beilstein Reference: 116316 MDL No.: MFCD00005776

Formula: C₇H₄ClNS
MW: 169.63 g/mol
Boiling Pt: 141 °C/30 mm
Melting Pt: 30…33 °C
Density: 1.370
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005776
CAS Number: 615-20-3
EINECS: 210-415-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR