m-Tolylacetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(m-Tolyl)acetic acid, (3-Methylphenyl)acetic acid

A17743-30 A17743-09 A17743-18
CAAAAA17743-30EA 835.82 CAD
CAAAAA17743-30 CAAAA17743-09 CAAAA17743-18
m-Tolylacetic acid 99%
m-Tolylacetic acid

Purity: 99% CAS No.: 621-36-3 Molecular Formula: C9H10O2 Formula Weight: 150.18 Melting Point: 62-66° Beilstein Reference: 2043549

Formula: (C₆H₄(CH₃))CH₂COOH
MW: 150.18 g/mol
Boiling Pt: 120…123 °C (26 mmHg)
Melting Pt: 62…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00004340
CAS Number: 621-36-3
EINECS: 210-683-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR