1,3-Dihydroxynaphthalene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Naphthalene-1,3-diol, Naphthalene-1,3-diol

A17739-03 A17739-06
CAAAA17739-03EA 71.57 CAD
CAAAA17739-03 CAAAA17739-06
1,3-Dihydroxynaphthalene 98%
1,3-Dihydroxynaphthalene

Purity: 98% CAS No.: 132-86-5 Molecular Formula: C10H8O2 Formula Weight: 160.17 Melting Point: 123-126° Beilstein Reference: 2044002 Merck Reference: 14,6396 MDL No.: MFCD00003965

Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Boiling Pt: 386 °C (1013 hPa)
Melting Pt: 123…126 °C
Density: 1.12 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003965
CAS Number: 132-86-5
EINECS: 205-079-7
Merck Index: 14,6396

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR