4,5-Dichloro-o-phenylenediamine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4,5-Dichloro-1,2-phenylenediamine, 1,2-Diamino-4,5-dichlorobenzene, 4,5-Dichlorobenzene-1,2-diamine

A17738-22 A17738-06 A17738-14
CAAAAA17738-22EA 1537.34 CAD
CAAAAA17738-22 CAAAA17738-06 CAAAA17738-14
4,5-Dichloro-o-phenylenediamine 98%
4,5-Dichloro-o-phenylenediamine

Purity: 98% CAS No.: 5348-42-5 Molecular Formula: C6H6Cl2N2 Formula Weight: 177.03 Melting Point: ca 162° dec. Beilstein Reference: 1072811 UN No.: 2811 MDL No.: MFCD00007723

Formula: C₆H₆Cl₂N₂
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: ∼ 162 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007723
CAS Number: 5348-42-5
EINECS: 226-305-0
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR