5-Nitro-o-toluidine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
5-Nitro-2-methylaniline, 2-Methyl-5-nitroaniline, 2-Amino-4-nitrotoluene
A17726-0E
A17726-22
A17726-36
CAAAA17726-0EEA
897.44
CAD
CAAAA17726-0E
CAAAA17726-22
CAAAA17726-36
5-Nitro-o-toluidine 98+%
5-Nitro-o-toluidine
Purity: 98+%
CAS No.: 99-55-8
Molecular Formula: C7H8N2O2
Formula Weight: 152.15
Odor: Characteristic
Melting Point: 105-108°
Beilstein Reference: 879021
UN No.: 2660
MDL No.: MFCD00007741
Formula:
C₇H₈N₂O₂ MW: 152.15 g/mol Melting Pt: 108 Storage Temperature: Ambient |
MDL Number:
MFCD00007741 CAS Number: 99-55-8 EINECS: 202-765-8 UN: 2660 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...