L(-)-Mannose 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
(2R,3R,4S,5S)-2,3,4,5,6-Pentahydroxyhexanal, L-(-)-Mannose
A17722-01
A17722-03
CAAAA17722-01EA
80.9
CAD
CAAAA17722-01
CAAAA17722-03
L(-)-Mannose 99%
L(-)-Mannose
Purity: 99%
CAS No.: 10030-80-5
Molecular Formula: C6H12O6
Formula Weight: 180.16
Melting Point: 129-131°
Beilstein Reference: 1724628
Formula:
C₆H₁₂O₆ MW: 180.16 g/mol Boiling Pt: 376 °C (1013 hPa) Melting Pt: 129…131 °C Storage Temperature: Ambient |
MDL Number:
MFCD00136021 CAS Number: 10030-80-5 EINECS: 233-080-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...