D-(+)-Tyrosine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17709-14
A17709-03
A17709-06
CAAAAA17709-14EA
606.72
CAD
CAAAAA17709-14
CAAAA17709-03
CAAAA17709-06
D-(+)-Tyrosine 99%
D(+)-Tyrosine
Purity: 99%
CAS No.: 556-02-5
Molecular Formula: C9H11NO3
Formula Weight: 181.19
Melting Point: 310-314°
Beilstein Reference: 2212157
Merck Reference: 14,9839
Optical Rotation: +11° (c=4 in 1N HCl)
MDL No.: MFCD00063073
Formula:
C₉H₁₁NO₃ Boiling Pt: 351 °C (1013 hPa) Melting Pt: 310…314 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063073 CAS Number: 556-02-5 EINECS: 209-112-6 Merck Index: 14,9839 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...