1-Bromo-4-nitrobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromonitrobenzene

A17705-06 A17705-14 A17705-22 A17705-36
CAAAAA17705-06EA 48.03 CAD
CAAAAA17705-06 CAAAA17705-14 CAAAA17705-22 CAAAA17705-36
1-Bromo-4-nitrobenzene 98%
1-Bromo-4-nitrobenzene

Purity: 98% CAS No.: 586-78-7 Molecular Formula: C6H4BrNO2 Formula Weight: 202.01 Melting Point: 124-127° Boiling Point: 254-256° UN No.: 3459 MDL No.: MFCD00007280

Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
Boiling Pt: 255
Melting Pt: 124…127 °C
Density: 1.948
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007280
CAS Number: 586-78-7
EINECS: 209-583-8
UN: 3459
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR