Heptanoic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A17704-0F
A17704-AP
CAAAA17704-0FEA
379.2
CAD
CAAAA17704-0F
CAAAA17704-AP
Heptanoic acid 98+%
Heptanoic acid
Purity: 98+%
CAS No.: 111-14-8
Molecular Formula: C7H14O2
Formula Weight: 130.19
Flash Point: 114°(237°F)
Refractive Index: 1.4230
Melting Point: -8°
Boiling Point: 222-223°
Beilstein Reference: 1744723
Merck Reference: 14,4660
UN No.: 3265
MDL No.: MFCD00004426
Formula:
C₇H₁₄O₂ Boiling Pt: 222…223 °C Melting Pt: –8 °C Density: 0.917 Flash Pt: 114 °C (237 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004426 CAS Number: 111-14-8 EINECS: 203-838-7 UN: 3265 ADR: 8,III Merck Index: 14,4660 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...