7-Isopropyl-1,4-dimethylazulene 98+%

Supplier: Thermo Scientific Chemicals
Warning

A17690-06 A17690-14
CAAAA17690-06EA 62.73 CAD
CAAAA17690-06 CAAAA17690-14
7-Isopropyl-1,4-dimethylazulene 98+%
7-Isopropyl-1,4-dimethylazulene

Purity: 98+% CAS No.: 489-84-9 Molecular Formula: C15H18 Formula Weight: 198.31 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 31-33° Boiling Point: 152-154°/7mm Beilstein Reference: 1365001 Merck Reference: 14,4554 MDL No.: MFCD00003811

Formula: C₁₅H₁₈
Boiling Pt: 152…154 °C/7 mm
Melting Pt: 31…33 °C
Density: 0.976
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00003811
CAS Number: 489-84-9
EINECS: 207-701-2
Merck Index: 14,4554

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR