1,3-Bis(diethylamino)propane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N1,N1,N3,N3-Tetraethylpropane-1,3-diamine, N,N,N',N'-Tetraethyl-1,3-diaminopropane, N,N,N',N'-Tetraethyltrimethylenediamine, N,N,N',N'-Tetraethyl-1,3-propanediamine
A17680-06
A17680-14
CAAAA17680-06EA
94.64
CAD
CAAAA17680-06
CAAAA17680-14
1,3-Bis(diethylamino)propane 97%
1,3-Bis(diethylamino)propane
Purity: 97%
CAS No.: 60558-96-5
Molecular Formula: (CH3CH2)2N(CH2)3N(CH2CH3)2
Formula Weight: 186.34
Flash Point: 76°(168°F)
Refractive Index: 1.4377
Boiling Point: 133°/80mm
Beilstein Reference: 1741257
UN No.: 2735
MDL No.: MFCD00009057
Formula:
C₁₁H₂₆N₂ MW: 186.34 Boiling Pt: 133 °C (80 mmHg) Density: 0.810 Flash Pt: 76 °C (168 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009057 CAS Number: 60558-96-5 UN: 2735 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...