Pentafluorobenzenesulfonyl chloride 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Pentafluorobenzene-1-sulfonyl chloride, 2,3,4,5,6-Pentafluorobenzenesulphonyl chloride, Pentafluorobenzene-1-sulphonyl chloride
A17676-03
A17676-06
A17676-14
CAAAAA17676-03EA
40.92
CAD
CAAAAA17676-03
CAAAA17676-06
CAAAA17676-14
Pentafluorobenzenesulfonyl chloride 98+%
Pentafluorobenzenesulphonyl chloride
Purity: 98+%
CAS No.: 832-53-1
Molecular Formula: C6ClF5O2S
Formula Weight: 266.57
Flash Point: 90°(194°F)
Refractive Index: 1.4790
Boiling Point: 210-211°
Beilstein Reference: 2142325
UN No.: 3265
MDL No.: MFCD00007427
Formula:
C₆ClF₅O₂S Boiling Pt: 210…211 °C Density: 1.796 Flash Pt: 90 °C (194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007427 CAS Number: 832-53-1 EINECS: 212-620-0 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...