Stearic acid 90+%
Supplier: Thermo Scientific Chemicals
A17673-0B
A17673-0I
CAAAA17673-0BEA
55.77
CAD
CAAAA17673-0B
CAAAA17673-0I
Stearic acid 90+%
Stearic acid
Purity: 90+%
CAS No.: 57-11-4
Molecular Formula: CH3(CH2)16CO2H
Formula Weight: 284.48
Flash Point: 196°(384°F)
Odor: Odorless
Boiling Point: 232°/15mm
Beilstein Reference: 608585
Merck Reference: 14,8804
MDL No.: MFCD00002752
Formula:
C₁₈H₃₆O₂ Boiling Pt: 232 °C/15 mm Melting Pt: 67…70 °C Density: 0.941 Flash Pt: 196 °C(384 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002752 CAS Number: 57-11-4 EINECS: 200-313-4 Merck Index: 14,8804 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...