α,3,4-Trichlorotoluene 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,2-Dichloro-4-(chloromethyl)benzene, 1-(Chloromethyl)-3,4-dichlorobenzene, 3,4-Dichlorobenzyl chloride

A17672-18 A17672-30 A17672-14 A17672-22
CAAAAA17672-18EA 73.15 CAD
CAAAAA17672-18 CAAAAA17672-30 CAAAA17672-14 CAAAA17672-22
α,3,4-Trichlorotoluene 98%
α,3,4-Trichlorotoluene

Purity: 98% CAS No.: 102-47-6 Molecular Formula: C7H5Cl3 Formula Weight: 195.48 Flash Point: 155°(311°F) Refractive Index: 1.5766 Odor: Pungent Melting Point: -3° Boiling Point: 99-101°/2mm Beilstein Reference: 386644 UN No.: 3265 MDL No.: MFCD00000906

Formula: C₇H₅Cl₃
MW: 195.47 g/mol
Boiling Pt: 99…101 °C/2 mm
Melting Pt: –3 °C
Density: 1.416
Flash Pt: 155 °C (311 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000906
CAS Number: 102-47-6
EINECS: 203-033-0
UN: 1760
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR