3-Nitrobenzenesulfonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

A17670-14 A17670-22
CAAAA17670-14EA 121.82 CAD
CAAAA17670-14 CAAAA17670-22
3-Nitrobenzenesulfonyl chloride 98%
3-Nitrobenzenesulphonyl chloride

Purity: 98% CAS No.: 121-51-7 Molecular Formula: C6H4ClNO4S Formula Weight: 221.62 Flash Point: 110°(230°F) Melting Point: 59-63° Beilstein Reference: 746542 UN No.: 3261 MDL No.: MFCD00007435

Formula: C₆H₄ClNO₄S
Melting Pt: 59…63 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007435
CAS Number: 121-51-7
EINECS: 204-476-2
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR