Oxybenzone 98+%
Supplier: Thermo Scientific Chemicals
A17662-0B
A17662-18
A17662-30
CAAAAA17662-0BEA
428.18
CAD
CAAAAA17662-0B
CAAAA17662-18
CAAAA17662-30
Oxybenzone 98+%
Oxybenzone
Purity: 98+%
CAS No.: 131-57-7
Molecular Formula: C14H12O3
Formula Weight: 228.25
Flash Point: 216°(420°F)
Melting Point: 62-65°
Boiling Point: ca 150°/5mm
Beilstein Reference: 1913145
Merck Reference: 14,6954
MDL No.: MFCD00008387
Formula:
C₁₄H₁₂O₃ Boiling Pt: ∼ 150 °C/5 mm Melting Pt: 62…65 °C Density: 1.300 Flash Pt: 216 °C (420 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008387 CAS Number: 131-57-7 EINECS: 205-031-5 Merck Index: 14,6954 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...