2-Undecanone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl nonyl ketone, Nonyl methyl ketone
A17659-0E
A17659-36
CAAAA17659-0EEA
886.38
CAD
CAAAA17659-0E
CAAAA17659-36
2-Undecanone 98%
2-Undecanone
Purity: 98%
CAS No.: 112-12-9
Molecular Formula: CH3(CH2)8COCH3
Formula Weight: 170.30
Flash Point: 89°(192°F)
Refractive Index: 1.4300
Melting Point: 11-13°
Boiling Point: 228°
Beilstein Reference: 1749573
Merck Reference: 14,6104
UN No.: 3082
MDL No.: MFCD00009583
Formula:
C₁₁H₂₂O MW: 170.3 g/mol Boiling Pt: 228 °C Melting Pt: 11…13 °C Density: 0.825 Flash Pt: 89 °C (192 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009583 CAS Number: 112-12-9 EINECS: 203-937-5 UN: 3082 ADR: 9,III Merck Index: 14,6104 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...