2-Methoxynaphthalene 98%
Supplier: Thermo Scientific Chemicals
A17650-0B
A17650-30
CAAAA17650-0BEA
205.4
CAD
CAAAA17650-0B
CAAAA17650-30
2-Methoxynaphthalene 98%
2-Methoxynaphthalene
Purity: 98%
CAS No.: 93-04-9
Molecular Formula: C11H10O
Formula Weight: 158.20
Flash Point: ca 139°(282°F)
Melting Point: 70-75°
Boiling Point: 272-274°
Beilstein Reference: 1859408
Merck Reference: 14,5997
MDL No.: MFCD00004061
Formula:
C₁₁H₁₀O Boiling Pt: 272…274 °C Melting Pt: 70…75 °C Density: 1.266 g/cm³ (20 °C) Flash Pt: ∼ 139 °C (282 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004061 CAS Number: 93-04-9 EINECS: 202-213-6 Merck Index: 14,5997 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...