(R)-(-)-Hydratropic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-(-)-2-Phenylpropanoic acid, (R)-(-)-2-Phenylpropionic acid, (R)-2-Phenylpropanoic acid

A17639-01 A17639-03
CAAAAA17639-06EA 975.38 CAD
CAAAAA17639-06 CAAAA17639-01 CAAAA17639-03
(R)-(-)-Hydratropic acid 97%
(R)-(-)-Hydratropic acid

Purity: 97% CAS No.: 7782-26-5 Molecular Formula: C9H10O2 Formula Weight: 150.18 Flash Point: 148°(298°F) Refractive Index: 1.5230 Melting Point: 32-34° Boiling Point: 264-266° Beilstein Reference: 2207688 Optical Rotation: -73° (c=1.6 in chloroform) MDL No.: MFCD00063140

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 264…266 °C
Melting Pt: 32…34 °C
Density: 1,100
Flash Pt: 148 °C(298 °F)
Storage Temperature: Ambient
MDL Number: MFCD00063140
CAS Number: 7782-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR