5-Bromo-α,α,α-trifluoro-2-nitrotoluene 98+%

Supplier: Thermo Scientific Chemicals
Warning

A17634-06 A17634-14 A17634-22
CAAAA17634-06EA 29.55 CAD
CAAAA17634-06 CAAAA17634-14 CAAAA17634-22
5-Bromo-α,α,α-trifluoro-2-nitrotoluene 98+%
5-Bromo-α,α,α-trifluoro-2-nitrotoluene

Purity: 98+% CAS No.: 344-38-7 Molecular Formula: C7H3BrF3NO2 Formula Weight: 270.01 Flash Point: 110°(230°F) Refractive Index: 1.5180 Melting Point: 32-35° Boiling Point: 95-96°/5mm Beilstein Reference: 2650701 MDL No.: MFCD00014706

Formula: C₇H₃BrF₃NO₂
Boiling Pt: 95…96 °C/5 mm
Melting Pt: 32…35 °C
Density: 1.799
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014706
CAS Number: 344-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR