Benzyl bromoacetate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Benzyl 2-bromoacetate, Bromoacetic benzyl ester
A17630-0B
A17630-18
A17630-30
CAAAA17630-0BEA
543.52
CAD
CAAAA17630-0B
CAAAA17630-18
CAAAA17630-30
Benzyl bromoacetate 97%
Benzyl bromoacetate
Purity: 97%
CAS No.: 5437-45-6
Molecular Formula: C6H5CH2CO2CH2Br
Formula Weight: 229.08
Flash Point:
Formula:
C₉H₉BrO₂ MW: 229.07 g/mol Boiling Pt: 157…159 °C/10 mm Density: 1.460 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000190 CAS Number: 5437-45-6 EINECS: 226-611-4 |
Learn more
![](/m-ca.vwr.com/en_CA/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...