L(+)-Histidine monohydrochloride monohydrate 99%
Supplier: Thermo Scientific Chemicals
A17627-36
A17627-18
A17627-30
CAAAAA17627-36EA
311.26
CAD
CAAAAA17627-36
CAAAA17627-18
CAAAA17627-30
L(+)-Histidine monohydrochloride monohydrate 99%
L(+)-Histidine monohydrochloride monohydrate
Purity: 99%
CAS No.: 5934-29-2
Molecular Formula: C6H9N3O2.HCl.H2O
Formula Weight: 209.63 (191.62anhy)
Melting Point: ca 240° dec.
Beilstein Reference: 4168261
Merck Reference: 14,4720
Optical Rotation: +10° (c=5 in 5N HCl)
MDL No.: MFCD00151027
Formula:
C₆H₉N₃O₂·HCl·H₂O Melting Pt: ∼ 240 °C (dec.) Density: 1.485 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00151027 CAS Number: 5934-29-2 EINECS: 211-438-9 Merck Index: 14,4720 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...