2-Bromethylamine hydrobromide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17625-0B
A17625-18
A17625-30
CAAAAA17625-0BEA
756.82
CAD
CAAAAA17625-0B
CAAAA17625-18
CAAAA17625-30
2-Bromethylamine hydrobromide 98+%
2-Bromethylamine hydrobromide
Purity: 98+%
CAS No.: 2576-47-8
Molecular Formula: BrCH2CH2NH2.HBr
Formula Weight: 204.90
Melting Point: 172-176°
Beilstein Reference: 3607605
Formula:
BrCH₂CH₂NH₃Br Melting Pt: 172…176 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012886 CAS Number: 2576-47-8 EINECS: 219-924-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...