4-Fluorothiophenol, (max. 7% H₂O) 98+% (dry weight)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Fluorobenzene-1-thiol, 1-Fluoro-4-mercaptobenzene, 4-Fluorobenzenethiol, p-Fluorothiophenol

A17600-06 A17600-14 A17600-22
CAAAA17600-06EA 86.74 CAD
CAAAA17600-06 CAAAA17600-14 CAAAA17600-22
4-Fluorothiophenol, (max. 7% H₂O) 98+% (dry weight)
4-Fluorothiophenol

Purity: 98+% CAS No.: 371-42-6 Molecular Formula: C6H5FS Formula Weight: 128.17 Flash Point: 57°(135°F) Refractive Index: 1.5500 Odor: Unpleasant Boiling Point: 170-172° Beilstein Reference: 1446350 UN No.: 3071 MDL No.: MFCD00004846

Formula: C₆H₅FS
MW: 128.17 g/mol
Boiling Pt: 170…172 °C
Density: 1.203
Flash Pt: 57 °C (135 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004846
CAS Number: 371-42-6
EINECS: 206-737-6
UN: 1228
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR