Dexamethasone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17590-03
A17590-06
A17590-14
CAAAA17590-03EA
205.4
CAD
CAAAA17590-03
CAAAA17590-06
CAAAA17590-14
Dexamethasone 98%
Dexamethasone
Anti-inflammatory glucocorticoid
Purity: 98%
CAS No.: 50-02-2
Molecular Formula: C22H29FO5
Formula Weight: 392.47
Melting Point: ca 260° dec.
Beilstein Reference: 2066651
Merck Reference: 14,2943
Optical Rotation: +75° (c=1 in doxane)
MDL No.: MFCD00064136
Anti-inflammatory glucocorticoid
Formula:
C₂₂H₂₉FO₅ Melting Pt: ca 260 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00064136 CAS Number: 50-02-2 EINECS: 200-003-9 Merck Index: 14,2943 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...