Butyryl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A17575-0B
A17575-30
CAAAA17575-0BEA
188.02
CAD
CAAAA17575-0B
CAAAA17575-30
Butyryl chloride 98%
Butyryl chloride
Purity: 98%
CAS No.: 141-75-3
Molecular Formula: CH3CH2CH2COCl
Formula Weight: 106.55
Flash Point: 18°(64°F)
Refractive Index: 1.4120
Odor: Acrid
Melting Point: -89°
Boiling Point: 101-102°
Beilstein Reference: 605395
Merck Reference: 14,1598
UN No.: 2353
MDL No.: MFCD00000752
Formula:
C₄H₇ClO Boiling Pt: 101…102 °C Melting Pt: –89 °C Density: 1.026 Flash Pt: 18 °C (64 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000752 CAS Number: 141-75-3 EINECS: 205-498-5 UN: 2353 ADR: 3,II Merck Index: 14,1598 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...