N-Acetyl-DL-tryptophan 99%
Supplier: Thermo Scientific Chemicals
A17562-14
A17562-22
CAAAA17562-14EA
119.13
CAD
CAAAA17562-14
CAAAA17562-22
N-Acetyl-DL-tryptophan 99%
N-Acetyl-DL-tryptophan
Purity: 99%
CAS No.: 87-32-1
Molecular Formula: C13H14N2O3
Formula Weight: 246.27
Melting Point: ca 205° dec.
Beilstein Reference: 89478
Formula:
C₁₃H₁₄N₂O₃ Melting Pt: ∼ 205 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00005644 CAS Number: 87-32-1 EINECS: 201-739-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...