1,4-Benzodioxane 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,3-Dihydrobenzo[b][1,4]dioxine, 2,3-Dihydro-1,4-benzodioxin, Benzo-1,4-dioxane, 1,4-Benzodioxan

A17558-09 A17558-18 A17558-30
CAAAA17558-09EA 52.3 CAD
CAAAA17558-09 CAAAA17558-18 CAAAA17558-30
1,4-Benzodioxane 98%
1,4-Benzodioxane

Purity: 98% CAS No.: 493-09-4 Molecular Formula: C8H8O2 Formula Weight: 136.15 Flash Point: 87°(188°F) Refractive Index: 1.5490 Boiling Point: 216° Beilstein Reference: 120846 MDL No.: MFCD00006821

Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 216 °C
Density: 1.170
Flash Pt: 87 °C(188 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006821
CAS Number: 493-09-4
EINECS: 207-775-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR