2-Fluoro-5-(trifluoromethyl)aniline 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-Amino-alpha,alpha,alpha,4-tetrafluorotoluene, α,α,α,6-Tetrafluoro-m-toluidine, 3-Amino-α,α,α,4-tetrafluorotoluene, 3-Amino-4-fluorobenzotrifluoride
A17556-03
A17556-06
A17556-14
CAAAA17556-03EA
34.92
CAD
CAAAA17556-03
CAAAA17556-06
CAAAA17556-14
2-Fluoro-5-(trifluoromethyl)aniline 97%
2-Fluoro-5-(trifluoromethyl)aniline
Purity: 97%
CAS No.: 535-52-4
Molecular Formula: C7H5F4N
Formula Weight: 179.12
Flash Point: 70°(158°F)
Refractive Index: 1.4610
Odor: Characteristic
Boiling Point: 210°
Beilstein Reference: 1950800
UN No.: 2810
MDL No.: MFCD00007653
Formula:
C₇H₅F₄N MW: 179.12 g/mol Boiling Pt: 210 °C Density: 1.378 Flash Pt: 70 °C (158 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007653 CAS Number: 535-52-4 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...