Polyethylene glycol 6,000

Supplier: Thermo Scientific Chemicals

Synonyms: PEG 6000PEG 6,000

A17541-30 A17541-30 A17541-0B A17541-0B A17541-0I A17541-0I
CAAAA17541-30EA 44.24 CAD
CAAAA17541-30 AAA17541-30 CAAAA17541-0B AAA17541-0I AAA17541-0B CAAAA17541-0I
Polyethylene glycol 6,000
Polyethylene glycol 6,000
Formula: H(OCH₂CH₂)nOH
MW: 6000 g/mol
Melting Pt: 60…63 °C
MDL Number: MFCD01779614
CAS Number: 25322-68-3
EINECS: 203-473-3
Merck Index: 14,7568

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR