trans,trans-1,4-Diphenyl-1,3-butadiene 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(1E,3E)-1,4-Diphenylbuta-1,3-diene
A17531-06
A17531-14
A17531-22
CAAAA17531-06EA
49.93
CAD
CAAAA17531-06
CAAAA17531-14
CAAAA17531-22
trans,trans-1,4-Diphenyl-1,3-butadiene 98+%
trans,trans-1,4-Diphenyl-1,3-butadiene
Purity: 98+%
CAS No.: 538-81-8
Molecular Formula: C16H14
Formula Weight: 206.29
Odor: Odorless
Melting Point: 151-154°
Boiling Point: 350°/720mm
Beilstein Reference: 1905937
Merck Reference: 14,3320
MDL No.: MFCD00004791
Formula:
C₁₆H₁₄ MW: 206.29 g/mol Boiling Pt: 350 °C/720 mm Melting Pt: 151…154 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004791 CAS Number: 538-81-8 EINECS: 212-952-6 Merck Index: 14,3320 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...