(+)-Cinchonine 98+% cont. up to 3% quinidine/dihydroquinidine and 3% quinine/dihydroquinine

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cinchonine, (S)-Quinolin-4-yl((1S,2R,4S,5R)-5-vinylquinuclidin-2-yl)methanol

A17523-14 A17523-22
CAAAA17523-14EA 62.73 CAD
CAAAA17523-14 CAAAA17523-22
(+)-Cinchonine 98+% cont. up to 3% quinidine/dihydroquinidine and 3% quinine/dihydroquinine
(+)-Cinchonine

Purity: 98+% CAS No.: 118-10-5 Molecular Formula: C19H22N2O Formula Weight: 294.39 Melting Point: 253-258° Beilstein Reference: 89689 Merck Reference: 14,2287 Optical Rotation: +225° (c=0.5 in ethanol) MDL No.: MFCD00064372

Formula: C₁₉H₂₂N₂O
MW: 294.4 g/mol
Melting Pt: 253…258 °C
Density: 1.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064372
CAS Number: 118-10-5
EINECS: 204-234-6
Merck Index: 14,2287

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR