DL-Arginine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Arginine, (±)-Arginine, H-DL-Arg-OH, H-Arg-OH

A17520-06 A17520-14
CAAAA17520-06EA 54.83 CAD
CAAAA17520-06 CAAAA17520-14
DL-Arginine 98%
DL-Arginine

Purity: 98% CAS No.: 7200-25-1 Molecular Formula: C6H14N4O2 Formula Weight: 174.20 Melting Point: ca 230° dec. Beilstein Reference: 1725411

Formula: C₆H₁₄N₄O₂
MW: 174.2
Melting Pt: ∼ 230 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00063117
CAS Number: 7200-25-1
EINECS: 230-571-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR