Ethyl methacrylate 98+% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ethyl 2-methylpropenoate, Methacrylic acid ethyl ester, Ethyl-2-methylprop-2-enoat, Ethyl-2-methylpropenoat, Ethyl 2-methylprop-2-enoate
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ) 10 -120 ppm
Ethyl methacrylate is used to prepare copolymers of EMA on waxy maize starch and hydroxypropylstarch, potential application of these copolymers are as excipients for compressed non-disintegrating matrix tablets. It is also used to study reactions of n-butyl acrylate and EMA with ozone in the gas phase.
Purity: 98+%
CAS No.: 97-63-2
Molecular Formula: CH2=C(CH3)CO2CH2CH3
Formula Weight: 114.15
Storage Temperature: 2°C to 8°C
Flash Point: 18°(64°F)
Refractive Index: 1.4140
Odor: Pungent
Melting Point: -60°
Boiling Point: 118-119°
Beilstein Reference: 471201
UN No.: 2277
MDL No.: MFCD00009161
Formula:
C₆H₁₀O₂ MW: 114.14 g/mol Boiling Pt: 118…119 °C Melting Pt: –60 °C Density: 0.917 Flash Pt: 18 °C (64 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00009161 CAS Number: 97-63-2 EINECS: 202-597-5 UN: 2277 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...