3-Chloro-4-hydroxybenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

A17515-06 A17515-14
CAAAA17515-06EA 165.9 CAD
CAAAA17515-06 CAAAA17515-14
3-Chloro-4-hydroxybenzaldehyde 98%
3-Chloro-4-hydroxybenzaldehyde

Purity: 98% CAS No.: 2420-16-8 Molecular Formula: C7H5ClO2 Formula Weight: 156.57 Melting Point: 128-132° MDL No.: MFCD00016981

Formula: C₇H₅ClO₂
MW: 156.57 g/mol
Boiling Pt: 251 °C (1013 hPa)
Melting Pt: 128…132 °C
Density: 1,0404 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016981
CAS Number: 2420-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR