Bromocresol green sodium salt

Supplier: Thermo Scientific Chemicals

Synonyms: Bromocresol green water soluble, Sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-2,2-dioxido-3H-benzo[d][1,2]oxathiol-3-yl)-3-methylphenolate

A17503-22 A17503-06 A17503-14
CAAAAA17503-22EA 704.68 CAD
CAAAAA17503-22 CAAAA17503-06 CAAAA17503-14
Bromocresol green sodium salt
Bromocresol green sodium salt

CAS No.: 62625-32-5 Molecular Formula: C21H13Br4NaO5S Formula Weight: 720.01 Melting Point: ca 230° dec. MDL No.: MFCD00148898

Formula: C₂₁H₁₃Br₄NaO₅S
MW: 720 g/mol
Melting Pt: 225 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00148898
CAS Number: 62625-32-5
EINECS: 263-657-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR