p-Toluenesulfonyl isocyanate 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tosyl isocyanate, 4-Isocyanatosulphonyltoluene, 4-Isocyanatosulfonyltoluene

A17492-14 A17492-22 A17492-36
CAAAAA17492-14EA 39.03 CAD
CAAAAA17492-14 CAAAA17492-22 CAAAA17492-36
p-Toluenesulfonyl isocyanate 95%
p-Toluenesulphonyl isocyanate

Purity: 95% CAS No.: 4083-64-1 Molecular Formula: C8H7NO3S Formula Weight: 197.21 Flash Point: 145°(293°F) Refractive Index: 1.5350 Odor: Irritating Melting Point: 5° Boiling Point: 144°/10mm Beilstein Reference: 391287 UN No.: 2206 MDL No.: MFCD00002030

Formula: C₈H₇NO₃S
Boiling Pt: 144 °C/10 mm
Melting Pt: 5 °C
Density: 1.295
Flash Pt: 145 °C(293 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002030
CAS Number: 4083-64-1
EINECS: 223-810-8
UN: 2206
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR